Ethoxytrimethylsilane, 97%, Thermo Scientific Chemicals
Ethoxytrimethylsilane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethoxytrimethylsilane, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
100 g
Catalog number L10151.22
also known as L10151-22
Price (USD)/ Each
111.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
111.00
Add to cart
Ethoxytrimethylsilane, 97%, Thermo Scientific Chemicals
Catalog numberL10151.22
Price (USD)/ Each
111.00
-
Add to cart
Chemical Identifiers
CAS1825-62-3
IUPAC Nameethoxytrimethylsilane
Molecular FormulaC5H14OSi
InChI KeyRSIHJDGMBDPTIM-UHFFFAOYSA-N
SMILESCCO[Si](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Identification (FTIR)Conforms
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.3715-1.3765 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only