Thermo Scientific Chemicals

4-Nitrophenyl-beta-D-glucopyranoside, 98+%, Thermo Scientific Chemicals

Catalog number: L10154.06
5 g, Each
Thermo Scientific Chemicals

4-Nitrophenyl-beta-D-glucopyranoside, 98+%, Thermo Scientific Chemicals

Catalog number: L10154.06
5 g, Each
Quantity
Catalog number: L10154.06
also known as L10154-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2492-87-7
IUPAC Name
(2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol
Molecular Formula
C12H15NO8
InChI Key
IFBHRQDFSNCLOZ-RMPHRYRLSA-N
SMILES
OC[C@H]1O[C@@H](OC2=CC=C(C=C2)[N+]([O-])=O)[C@H](O)[C@@H](O)[C@@H]1O
Appearance (Color)
White to cream to yellow
Form
Powder
Assay (HPLC)
≥98.0%
Identification (FTIR)
Conforms
Melting Point
160-170°C

Description

4-Nitrophenyl-β-D-glucopyranoside acts as a chromogenic beta-D-glucosidase substrate used for the measurement of beta-glucosidase activity. Further, it is used in research, biochemical enzyme assays and in vitro diagnostic analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrophenyl-β-D-glucopyranoside acts as a chromogenic beta-D-glucosidase substrate used for the measurement of beta-glucosidase activity. Further, it is used in research, biochemical enzyme assays and in vitro diagnostic analysis.

Solubility
Soluble in water, methanol, warm ethanol, dimethyl sulfoxide and dimethylformamide. Insoluble in ether.

Notes
Store in a cool place. Incompatible with strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text