5,6,7,8-Tetrahydro-2-naphthol, 98%, Thermo Scientific Chemicals
5,6,7,8-Tetrahydro-2-naphthol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5,6,7,8-Tetrahydro-2-naphthol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L10164.06
also known as L10164-06
Price (USD)/ Each
39.30
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
39.30
Add to cart
5,6,7,8-Tetrahydro-2-naphthol, 98%, Thermo Scientific Chemicals
Catalog numberL10164.06
Price (USD)/ Each
39.30
-
Add to cart
Chemical Identifiers
CAS1125-78-6
IUPAC Name5,6,7,8-tetrahydronaphthalen-2-ol
Molecular FormulaC10H12O
InChI KeyUMKXSOXZAXIOPJ-UHFFFAOYSA-N
SMILESOC1=CC=C2CCCCC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)57.5-63.5?C
Appearance (Color)White to cream
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only