3,5-Dimethylphenylacetonitrile, 98%, Thermo Scientific Chemicals
3,5-Dimethylphenylacetonitrile, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,5-Dimethylphenylacetonitrile, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number L10170.04
also known as L10170-04
Price (USD)/ Each
77.30
-
Add to cart
Quantity:
2 g
Request bulk or custom format
Price (USD)/ Each
77.30
Add to cart
3,5-Dimethylphenylacetonitrile, 98%, Thermo Scientific Chemicals
Catalog numberL10170.04
Price (USD)/ Each
77.30
-
Add to cart
Chemical Identifiers
CAS39101-54-7
IUPAC Name2-(3,5-dimethylphenyl)acetonitrile
Molecular FormulaC10H11N
InChI KeyLMUKNQSVBFEUKR-UHFFFAOYSA-N
SMILESCC1=CC(CC#N)=CC(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)Colorless to white to cream to pale yellow
Identification (FTIR)Conforms
FormCrystals or powder or crystalline powder or lumps or fused solid or liquid as melt
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only