Triethyl 3-phosphonopropionate, 98%, Thermo Scientific Chemicals
Triethyl 3-phosphonopropionate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Triethyl 3-phosphonopropionate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number L10190.09
also known as L10190-09
Price (USD)/ Each
58.30
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
58.30
Add to cart
Triethyl 3-phosphonopropionate, 98%, Thermo Scientific Chemicals
Catalog numberL10190.09
Price (USD)/ Each
58.30
-
Add to cart
Chemical Identifiers
CAS3699-67-0
IUPAC Nameethyl 3-(diethoxyphosphoryl)propanoate
Molecular FormulaC9H19O5P
InChI KeyJMJWCUOIOKBVNQ-UHFFFAOYSA-N
SMILESCCOC(=O)CCP(=O)(OCC)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4305-1.4345 @ 20?C
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only