Thermo Scientific Chemicals

10,12-Tricosadiynoic acid, 96%, Thermo Scientific Chemicals

Catalog number: L10231.03
1 g, Each
Thermo Scientific Chemicals

10,12-Tricosadiynoic acid, 96%, Thermo Scientific Chemicals

Catalog number: L10231.03
1 g, Each
Quantity
Catalog number: L10231.03
also known as L10231-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
66990-30-5
Form
Crystalline powder or crystals
Melting Point
55-60°C
Assay (Aqueous acid-base Titration)
≥95.0 to ≤105.0%
Appearance (Color)
White to blue

Description

It is an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text