5-Amino-2-methoxyphenol, 98%, Thermo Scientific Chemicals
5-Amino-2-methoxyphenol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Amino-2-methoxyphenol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L10234.06
also known as L10234-06
Price (USD)/ Each
74.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
74.00
Add to cart
5-Amino-2-methoxyphenol, 98%, Thermo Scientific Chemicals
Catalog numberL10234.06
Price (USD)/ Each
74.00
-
Add to cart
Chemical Identifiers
CAS1687-53-2
IUPAC Name5-amino-2-methoxyphenol
Molecular FormulaC7H9NO2
InChI KeyBLQFHJKRTDIZLX-UHFFFAOYSA-N
SMILESCOC1=CC=C(N)C=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Melting Point129-135?C
Appearance (Color)Brown to dark brown
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only