1-Cyclohexene-1-carbonitrile has been used in the preparation of cyclohexene-1-carboxylic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Cyclohexene-1-carbonitrile has been used in the preparation of cyclohexene-1-carboxylic acid.
Solubility
Not miscible in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
- R.L.Cargill; D.F.Bushey; J.J.Good. Alkylation of 1-cyanocyclohexene. J. Org. Chem. 1979, 44, (2),300-301
- Kiyoshi Iwai; Henry J.Shine. Ion radicals. 46. Reactions of the adducts of thianthrene and phenoxathiin cation radicals and cyclohexene with nucleophiles. J. Org. Chem. 1981, 46, (2),271-276