Ethyl 4,4,4-trifluorocrotonate, 98%, Thermo Scientific Chemicals
Ethyl 4,4,4-trifluorocrotonate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 4,4,4-trifluorocrotonate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L10285.03
also known as L10285-03
Price (USD)/ Each
31.10
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
31.10
Add to cart
Ethyl 4,4,4-trifluorocrotonate, 98%, Thermo Scientific Chemicals
Catalog numberL10285.03
Price (USD)/ Each
31.10
-
Add to cart
Chemical Identifiers
CAS25597-16-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only