Thermo Scientific Chemicals

Tantalum(V) ethoxide, 99+%, Thermo Scientific Chemicals

Catalog number: L10288.06
5 g, Each
Thermo Scientific Chemicals

Tantalum(V) ethoxide, 99+%, Thermo Scientific Chemicals

Catalog number: L10288.06
5 g, Each
Quantity
Catalog number: L10288.06
also known as L10288-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6074-84-6
IUPAC Name
pentaethoxytantalum
Molecular Formula
C10H25O5Ta
InChI Key
HSXKFDGTKKAEHL-UHFFFAOYSA-N
SMILES
CCO[Ta](OCC)(OCC)(OCC)OCC
Form
Liquid (as melt)
Elemental Analysis (typical)
Magnesium, Mg <20 ppm
Elemental Analysis (typical)
Sodium, Na <20 ppm
Elemental Analysis (typical)
Barium, Ba: <20 ppm
Refractive Index
1.4865-1.4910 (as melt)

Description

Tantalum(V) ethoxide acts as a precursor used in the preparation of ultra thin films of tantalum oxide and other tantalum containing films, which finds application in semiconductors. It is also used to prepare tantalum oxide nanoparticles for the imaging cartilage.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tantalum(V) ethoxide acts as a precursor used in the preparation of ultra thin films of tantalum oxide and other tantalum containing films, which finds application in semiconductors. It is also used to prepare tantalum oxide nanoparticles for the imaging cartilage.

Solubility
Soluble in organic solvents.

Notes
Moisture sensitive and air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text