Thermo Scientific Chemicals

(1-Tetradecyl)trimethylammonium bromide, 98%, Thermo Scientific Chemicals

Catalog number: L10294.36
500 g, Each
Thermo Scientific Chemicals

(1-Tetradecyl)trimethylammonium bromide, 98%, Thermo Scientific Chemicals

Catalog number: L10294.36
500 g, Each
Quantity
Catalog number: L10294.36
also known as L10294-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1119-97-7
IUPAC Name
trimethyl(tetradecyl)azanium bromide
Molecular Formula
C17H38BrN
InChI Key
CXRFDZFCGOPDTD-UHFFFAOYSA-M
SMILES
[Br-].CCCCCCCCCCCCCC[N+](C)(C)C
Appearance (Color)
White
Assay (Titration ex Bromide)
≥97.5 to ≤102.5%
Form
Crystals or powder or crystalline powder

Description

Myristyltrimethylammonium has been used in a study to assess a surfactant-controlled synthetic method to obtain a nanophase of mesoporous ceria-zirconia solid solution containing cationic defects in the crystalline structure.  It has also been used in a study to determine the low molecular weight of organic acids in soils and plants by capillary zone electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Myristyltrimethylammonium has been used in a study to assess a surfactant-controlled synthetic method to obtain a nanophase of mesoporous ceria-zirconia solid solution containing cationic defects in the crystalline structure.  It has also been used in a study to determine the low molecular weight of organic acids in soils and plants by capillary zone electrophoresis.

Solubility
Soluble in water. (100 g/L)

Notes
Light sensitive. Store in cool dry condition. Incompatible with strong oxidizing agents. Ensure ventilation.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text