Crystals or powder or crystalline powder or lumps or fused solid or granules
Appearance (Color)
Cream to yellow to dark green to green to brown or pale purple to pale gray to gray to black
Assay (HPLC)
≥98.0%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.