Thermo Scientific Chemicals

(R)-(+)-1-Phenylethyl isocyanate, 99%, Thermo Scientific Chemicals

Catalog number: L10305.06
5 g, Each
Thermo Scientific Chemicals

(R)-(+)-1-Phenylethyl isocyanate, 99%, Thermo Scientific Chemicals

Catalog number: L10305.06
5 g, Each
Quantity
Catalog number: L10305.06
also known as L10305-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
33375-06-3
Assay (GC)
≥98.5%
Appearance (Color)
Clear to slightly hazy, colorless
Form
Liquid
Refractive Index
1.5110-1.5150 @ 20?C
Optical Rotation
+10.5? ? 1.0? (neat)

Description

(R)-1-phenylethyl isocyanate is used in the derivatisation of alcohols, thiocarbamates and hydroxy fatty acids. (R)-1-phenylethyl isocyanate moderately inhibited both BP-DNA adducts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-1-phenylethyl isocyanate is used in the derivatisation of alcohols, thiocarbamates and hydroxy fatty acids. (R)-1-phenylethyl isocyanate moderately inhibited both BP-DNA adducts.

Solubility
Decomposes in water.

Notes
Moisture Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture. Store at 4°C.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text