Thermo Scientific Chemicals

trans-3-Hexene, 98%, Thermo Scientific Chemicals

Catalog number: L10311.06
5 g, Each
Thermo Scientific Chemicals

trans-3-Hexene, 98%, Thermo Scientific Chemicals

Catalog number: L10311.06
5 g, Each
Quantity
Catalog number: L10311.06
also known as L10311-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13269-52-8
IUPAC Name
(3E)-hex-3-ene
Molecular Formula
C6H12
InChI Key
ZQDPJFUHLCOCRG-AATRIKPKSA-N
SMILES
CC\C=C\CC
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
≥97.5% (non-U.S. sourced material)
Assay from Supplier's CofA
≥97.5% (U.S. specification)
Identification (FTIR)
Conforms

Description

Oligomers were prepared from the ozonolyses of cis- and trans-3-hexene and cis- and trans-2,5-dimethyl-3-hexene. Synthesis and characterization of aliphatic unsaturated polyesters is from trans-3-hexene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Oligomers were prepared from the ozonolyses of cis- and trans-3-hexene and cis- and trans-2,5-dimethyl-3-hexene. Synthesis and characterization of aliphatic unsaturated polyesters is from trans-3-hexene.

Solubility
Insoluble in water. Soluble in benzene, ligroin, ethanol, chloroform, ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text