Thermo Scientific Chemicals

1,3-Cyclopentanedione, 98%, Thermo Scientific Chemicals

Catalog number: L10336.03
1 g, Each
Thermo Scientific Chemicals

1,3-Cyclopentanedione, 98%, Thermo Scientific Chemicals

Catalog number: L10336.03
1 g, Each
Quantity
Catalog number: L10336.03
also known as L10336-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3859-41-4
IUPAC Name
cyclopentane-1,3-dione
Molecular Formula
C5H6O2
InChI Key
LOGSONSNCYTHPS-UHFFFAOYSA-N
SMILES
O=C1CCC(=O)C1
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥97.5%
Appearance (Color)
Cream to yellow to brown
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%

Description

1,3 cyclopentanedione is used in the synthesis of pgd2 . Reatant in the synthesis of fused dihydrofuran.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3 cyclopentanedione is used in the synthesis of pgd2 . Reatant in the synthesis of fused dihydrofuran.

Solubility
Very soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text