Thermo Scientific Chemicals

2,5-Dibromo-3,4-dinitrothiophene, 95%, Thermo Scientific Chemicals

Catalog number: L10348.03
1 g, Each
Thermo Scientific Chemicals

2,5-Dibromo-3,4-dinitrothiophene, 95%, Thermo Scientific Chemicals

Catalog number: L10348.03
1 g, Each
Quantity
Catalog number: L10348.03
also known as L10348-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
52431-30-8
Assay (GC)
≥94.0%
Form
Crystals or powder or crystalline powder or lumps or fused solid
Melting Point (clear melt)
129.0-142.0?C
Appearance (Color)
White to cream to yellow

Description

Reatant in the synthesis of bromodinitro compound, polymers via stille coupling. 3,4-diaminothiophene was obtained from the reduction of 2,5-dibromo-3,4-dinitrothiophene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reatant in the synthesis of bromodinitro compound, polymers via stille coupling. 3,4-diaminothiophene was obtained from the reduction of 2,5-dibromo-3,4-dinitrothiophene.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text