The LC3 Antibody Kit for Autophagy includes a rabbit polyclonal antibody against LC3B that has been validated for use in fluorescence microscopy and high content imaging and analysis. Chloroquine diphosphate is also included - following treatment with this compound, normal autophagic flux is disrupted and autophagosomes accumulate as a result of the disruption in normal autophagic flux.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection Method
Fluorescent
For Use With (Equipment)
Confocal Microscope, Fluorescence Microscope, High Content Analysis Instrument
Product Type
Dye
Dye Type
Unlabeled
Format
96-well plate, Slide(s)
Shipping Condition
Dry Ice
Contents & Storage
Kit includes includes a rabbit polyclonal antibody against LC3B and chloroquine diphosphate.