Thermo Scientific Chemicals

2,2,3,3,4,4,4-Heptafluorobutyl acrylate, 97%, stab. with 100 ppm 4-methoxyphenol, Thermo Scientific Chemicals

Catalog number: L10400.06
5 g, Each
Thermo Scientific Chemicals

2,2,3,3,4,4,4-Heptafluorobutyl acrylate, 97%, stab. with 100 ppm 4-methoxyphenol, Thermo Scientific Chemicals

Catalog number: L10400.06
5 g, Each
Quantity
Catalog number: L10400.06
also known as L10400-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
424-64-6
IUPAC Name
2,2,3,3,4,4,4-heptafluorobutyl prop-2-enoate
Molecular Formula
C7H5F7O2
InChI Key
PLXOUIVCSUBZIX-UHFFFAOYSA-N
SMILES
FC(F)(F)C(F)(F)C(F)(F)COC(=O)C=C
Refractive Index
1.3295-1.3345 @ 20?C
Form
Liquid
Solution Test
(10% w/v in diethyl ether) : Clear
Solution Test
(10% w/v in methanol) : Clear
Appearance (Color)
Clear, colorless

Description

2,2,3,3,4,4,4-Heptafluorobutyl acrylate is a useful fluorinated acrylate for proteomics research. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,3,3,4,4,4-Heptafluorobutyl acrylate is a useful fluorinated acrylate for proteomics research. It is also used as pharmaceutical intermediate.

Solubility
Not miscible in water.

Notes
Light sensitive. Incompatible with oxidizing agents, heat, ultraviolet radiation and free radical initiators.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text