2,2,3,3,4,4,4-Heptafluorobutyl acrylate is a useful fluorinated acrylate for proteomics research. It is also used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,2,3,3,4,4,4-Heptafluorobutyl acrylate is a useful fluorinated acrylate for proteomics research. It is also used as pharmaceutical intermediate.
Solubility
Not miscible in water.
Notes
Light sensitive. Incompatible with oxidizing agents, heat, ultraviolet radiation and free radical initiators.
RUO – Research Use Only
Bishnu P.Koiry; M.Moukwa; Nikhil K.Singha. Reversible addition-fragmentation chain transfer (RAFT) polymerization of 2,2,3,3,4,4,4-heptafluorobutyl acrylate (HFBA). Journal of Fluorine Chemistry. 2013, 153, 137-142
Peng Wang; K.L.Tan; C.C Ho; M.C.Khew; E.T.Kang. Surface modification of natural rubber latex films by graft copolymerization. European Polymer Journal. 2000, 36, (7),1323-1331