Thermo Scientific Chemicals

(S)-(+)-2-Hexanol, 98%, Thermo Scientific Chemicals

Catalog number: L10401.03
1 g, Each
Thermo Scientific Chemicals

(S)-(+)-2-Hexanol, 98%, Thermo Scientific Chemicals

Catalog number: L10401.03
1 g, Each
Quantity
Catalog number: L10401.03
also known as L10401-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
52019-78-0
IUPAC Name
hexan-2-ol
Molecular Formula
C6H14O
InChI Key
QNVRIHYSUZMSGM-UHFFFAOYNA-N
SMILES
CCCCC(C)O
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless
Optical Rotation
11.9 ± 0.1? (neat)
Form
Liquid
Refractive Index
1.4130-1.4170 @ 20?C

Description

S)-(+)-2-Hexanol is the S enantiomer used in the preparation of key intermediates for model studies in the total synthesis of antivirally active glycolipid cycloviracin B1.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
S)-(+)-2-Hexanol is the S enantiomer used in the preparation of key intermediates for model studies in the total synthesis of antivirally active glycolipid cycloviracin B1.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text