S)-(+)-2-Hexanol is the S enantiomer used in the preparation of key intermediates for model studies in the total synthesis of antivirally active glycolipid cycloviracin B1.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
S)-(+)-2-Hexanol is the S enantiomer used in the preparation of key intermediates for model studies in the total synthesis of antivirally active glycolipid cycloviracin B1.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
Amit Kumar Tyagi et. al. Antimicrobial Potential and Chemical Characterization of Serbian Liverwort (Porella arboris-vitae): SEM and TEM Observations.. Evidence-based Complementary and Alternative Medicine,. 2013, 2013 382927.