Hydriodic acid, 57% w/w aqueous solution stabilized with 1.5% hypophosphorus acid, Thermo Scientific™
Hydriodic acid, 57% w/w aqueous solution stabilized with 1.5% hypophosphorus acid, Thermo Scientific™
Thermo Scientific™

Hydriodic acid, 57% w/w aqueous solution stabilized with 1.5% hypophosphorus acid, Thermo Scientific™

 
Have Questions?
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number L10410.22
Price (USD)/ Each
44.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
52.80 
Save 8.15 (15%)
-
Add to cart
Request bulk or custom format
Price (USD)/ Each
44.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
52.80 
Save 8.15 (15%)
Add to cart
Hydriodic acid, 57% w/w aqueous solution stabilized with 1.5% hypophosphorus acid, Thermo Scientific™
Catalog numberL10410.22
Price (USD)/ Each
44.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
52.80 
Save 8.15 (15%)
-
Add to cart
Chemical Identifiers
CAS10034-85-2, 7732-18-5, 6303-21-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to orange to red-brown
FormLiquid
Assay (Aqueous acid-base Titration)≥55.0%
CommentMay darken on storage
Stabilizer0.75-1.5% H3PO2
Hydriodic acid is used to convert primary alcohol into alkyl iodide. It is used in organic and inorganic synthesis as one of the primary sources of iodine as well as a reducing agent. It is also used to cleave the ethers to get alkyl iodides and alcohols.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

RUO – Research Use Only