Thermo Scientific Chemicals

2,6-Dichlorophenyl isocyanate, 98%, Thermo Scientific Chemicals

Catalog number: L10421.06
5 g, Each
Thermo Scientific Chemicals

2,6-Dichlorophenyl isocyanate, 98%, Thermo Scientific Chemicals

Catalog number: L10421.06
5 g, Each
Quantity
Catalog number: L10421.06
also known as L10421-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
39920-37-1
IUPAC Name
1,3-dichloro-2-isocyanatobenzene
Molecular Formula
C7H3Cl2NO
InChI Key
HMVKMAMIRAVXAN-UHFFFAOYSA-N
SMILES
ClC1=CC=CC(Cl)=C1N=C=O
Appearance (Color)
White to cream
Melting Point (clear melt)
43-47?C
Form
Crystals or fused solid
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text