Thermo Scientific Chemicals

4-Isopropylbenzenesulfonyl chloride, 96%, Thermo Scientific Chemicals

Catalog number: L10428.06
5 g, Each
Thermo Scientific Chemicals

4-Isopropylbenzenesulfonyl chloride, 96%, Thermo Scientific Chemicals

Catalog number: L10428.06
5 g, Each
Quantity
Catalog number: L10428.06
also known as L10428-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
54997-90-9
IUPAC Name
4-(propan-2-yl)benzene-1-sulfonyl chloride
Molecular Formula
C9H11ClO2S
InChI Key
CETRNHJIXGITKR-UHFFFAOYSA-N
SMILES
CC(C)C1=CC=C(C=C1)S(Cl)(=O)=O
Assay (GC)
≥95.0%
Refractive Index
1.5360-1.5420 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Identification (FTIR)
Conforms
Form
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text