4-Isopropylbenzenesulfonyl chloride, 96%, Thermo Scientific Chemicals
4-Isopropylbenzenesulfonyl chloride, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Isopropylbenzenesulfonyl chloride, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number L10428.14
also known as L10428-14
Price (USD)/ Each
153.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
153.00
Add to cart
4-Isopropylbenzenesulfonyl chloride, 96%, Thermo Scientific Chemicals
Catalog numberL10428.14
Price (USD)/ Each
153.00
-
Add to cart
Chemical Identifiers
CAS54997-90-9
IUPAC Name4-(propan-2-yl)benzene-1-sulfonyl chloride
Molecular FormulaC9H11ClO2S
InChI KeyCETRNHJIXGITKR-UHFFFAOYSA-N
SMILESCC(C)C1=CC=C(C=C1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥95.0%
Refractive Index1.5360-1.5420 @ 20?C
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only