Thermo Scientific Chemicals

3-Amino-2,2-dimethyl-1-propanol, 95%, Thermo Scientific Chemicals

Catalog number: L10429.06
5 g, Each
Thermo Scientific Chemicals

3-Amino-2,2-dimethyl-1-propanol, 95%, Thermo Scientific Chemicals

Catalog number: L10429.06
5 g, Each
Quantity
Catalog number: L10429.06
also known as L10429-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
26734-09-8
IUPAC Name
3-amino-2,2-dimethylpropan-1-ol
Molecular Formula
C5H13NO
InChI Key
FNVOFDGAASRDQY-UHFFFAOYSA-N
SMILES
CC(C)(CN)CO
Appearance (Color)
Colorless to white
Water Content (Karl Fischer Titration)
≤2.5%
Form
Crystals or powder or crystalline powder or fused solid or waxy solid
Assay (GC)
≥94.0%

Description

Reatant in the synthesis of cyclic carbamates, cryptophycin analogues. Neopentanolamine with ammonia and hydrogen yeilds neopentyldiamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reatant in the synthesis of cyclic carbamates, cryptophycin analogues. Neopentanolamine with ammonia and hydrogen yeilds neopentyldiamine.

Solubility
Soluble in Dichloromethane, Chloroforn, Ether and Methanol

Notes
Hygroscopic,Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text