Thermo Scientific Chemicals

Triethyl 4-phosphonobutyrate, 97%, Thermo Scientific Chemicals

Catalog number: L10488.09
10 g, Each
Thermo Scientific Chemicals

Triethyl 4-phosphonobutyrate, 97%, Thermo Scientific Chemicals

Catalog number: L10488.09
10 g, Each
Quantity
Catalog number: L10488.09
also known as L10488-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2327-69-7
IUPAC Name
ethyl 4-(diethoxyphosphoryl)butanoate
Molecular Formula
C10H21O5P
InChI Key
FYESTJSRASJZOY-UHFFFAOYSA-N
SMILES
CCOC(=O)CCCP(=O)(OCC)OCC
Appearance (Color)
Clear colorless
Assay (GC)
≥96.0%
Form
Liquid
Refractive Index
1.4350-1.4400 @ 20?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text