Ethyl pentafluorobenzoate, 98%, Thermo Scientific Chemicals
Ethyl pentafluorobenzoate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl pentafluorobenzoate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L10523.06
also known as L10523-06
Price (USD)/ Each
62.80
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
62.80
Add to cart
Ethyl pentafluorobenzoate, 98%, Thermo Scientific Chemicals
Catalog numberL10523.06
Price (USD)/ Each
62.80
-
Add to cart
Chemical Identifiers
CAS4522-93-4
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥97.5%
Appearance (Color)Clear colorless to yellow
Refractive Index1.4270-1.4310 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only