2-Chlorobenzyl isothiocyanate, 97%, Thermo Scientific Chemicals
2-Chlorobenzyl isothiocyanate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Chlorobenzyl isothiocyanate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L10546.03
also known as L10546-03
Price (USD)/ Each
52.90
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
52.90
Add to cart
2-Chlorobenzyl isothiocyanate, 97%, Thermo Scientific Chemicals
Catalog numberL10546.03
Price (USD)/ Each
52.90
-
Add to cart
Chemical Identifiers
CAS18967-44-7
IUPAC Name1-chloro-2-(isothiocyanatomethyl)benzene
Molecular FormulaC8H6ClNS
InChI KeyRMVDNJDSLXQPAV-UHFFFAOYSA-N
SMILESClC1=CC=CC=C1CN=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Refractive Index1.6115-1.6175 @ 20?C
Appearance (Color)Clear pale yellow to yellow to orange to red
FormLiquid
2-Chlorobenzyl isothiocyanate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorobenzyl isothiocyanate is used as pharmaceutical intermediate.

Solubility
It hydrolyzes in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents, alcohols and amines.
RUO – Research Use Only

General References:

  1. Cliff C.Cheng; Gerald W.Shipps Jr.; Zhiwei Yang; Binyuan Sun; Noriyuki Kawahata; Kyle A.Soucy; Aileen Soriano; Peter Orth; Li Xiao; Paul Mann; Todd Black. Discovery and optimization of antibacterial AccC inhibitors. Bioorganic & Medicinal Chemistry Letters. 2009, 19, (23),6507-6514