Thermo Scientific Chemicals

2-Acetyl-3-aminothiophene, 97%, Thermo Scientific Chemicals

Catalog number: L10563.03
1 g, Each
Thermo Scientific Chemicals

2-Acetyl-3-aminothiophene, 97%, Thermo Scientific Chemicals

Catalog number: L10563.03
1 g, Each
Quantity
Catalog number: L10563.03
also known as L10563-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
31968-33-9
IUPAC Name
1-(3-aminothiophen-2-yl)ethan-1-one
Molecular Formula
C6H7NOS
InChI Key
MMLSKDGCLMDHAJ-UHFFFAOYSA-N
SMILES
CC(=O)C1=C(N)C=CS1
Form
Crystals or powder or crystalline powder or fused solid
Appearance (Color)
White to dark cream to cream to yellow to pale brown to gray
Assay (GC)
≥96.0%
Melting Point (clear melt)
84.0-93.0?C

Description

Dithienopyridines was synthesised from 2-Acetyl-3-aminothiophene. Also used as fine Intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dithienopyridines was synthesised from 2-Acetyl-3-aminothiophene. Also used as fine Intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text