Thermo Scientific Chemicals

Vitamin K{1}, Thermo Scientific Chemicals

Catalog number: L10575.03
1 g, Each
Thermo Scientific Chemicals

Vitamin K{1}, Thermo Scientific Chemicals

Catalog number: L10575.03
1 g, Each
Quantity
Catalog number: L10575.03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
84-80-0
IUPAC Name
2-methyl-3-[(2E)-3,7,11,15-tetramethylhexadec-2-en-1-yl]-1,4-dihydronaphthalene-1,4-dione
Molecular Formula
C31H46O2
InChI Key
MBWXNTAXLNYFJB-LKUDQCMENA-N
SMILES
CC(C)CCCC(C)CCCC(C)CCCC(C)=CCC1=C(C)C(=O)C2=CC=CC=C2C1=O
Appearance (Color)
Yellow or orange
Form
Viscous liquid
Assay from Supplier's CofA
≥97.5% (cis + trans)
Comment
5-25% cis isomer
Identification (FTIR)
Conforms

Description

Vitamin K{1} is an important dietary source found in green plants, algae and photosynthetic bacteria. It is used as a vitamin (prothrombogenic) and veterinary antidote for dicoumarol poisoning. It is an essential vitamin for the synthesis of coagulation factors II, VII, IX, and X in the liver.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text