Reagent for determination of serum iron. Reagent for the simultaneous spectrophotometric determination of iron and copper. PDT is the non-sulfonated precursor to Ferrozine (FZ), a commonly used reagent for Fe(II).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Reagent for determination of serum iron. Reagent for the simultaneous spectrophotometric determination of iron and copper. PDT is the non-sulfonated precursor to Ferrozine (FZ), a commonly used reagent for Fe(II).
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
R. E. Kitson. Simultaneous Spectrophotometric Determination of Cobalt, Copper, and Iron. Journal name. 1950, 22 (5), 664-667.
Peter L Croot,; Keith A Hunter.R. E. Kitson. Analytica Chimica Acta. 2000, 406 (2), 289-302.
Reagent for determination of serum iron: Anal. Chem., 42, 779 (1970).