Thermo Scientific Chemicals

2-Isopropylphenyl isocyanate, 97%, Thermo Scientific Chemicals

Catalog number: L10618.06
5 g, Each
Thermo Scientific Chemicals

2-Isopropylphenyl isocyanate, 97%, Thermo Scientific Chemicals

Catalog number: L10618.06
5 g, Each
Quantity
Catalog number: L10618.06
also known as L10618-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
56309-56-9
IUPAC Name
1-isocyanato-2-(propan-2-yl)benzene
Molecular Formula
C10H11NO
InChI Key
MWBSQVPLYJZPTG-UHFFFAOYSA-N
SMILES
CC(C)C1=CC=CC=C1N=C=O
Refractive Index
1.5215-1.5265 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥96.0%
Form
Liquid

Description

2-Isopropylphenyl isocyanate is used as intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Isopropylphenyl isocyanate is used as intermediates.

Solubility
Insoluble in water.

Notes
Moisture Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from moisture, alocohol, amines, water.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text