2-Isopropylphenyl isocyanate is used as intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Isopropylphenyl isocyanate is used as intermediates.
Solubility
Insoluble in water.
Notes
Moisture Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from moisture, alocohol, amines, water.
RUO – Research Use Only
General References:
- Hiroyuki Kai,; Yasuhide Moriok,; Takami Murashi,; Koichi Morita,; Satomi Shinonome,; Hitoshi Nakazato. 2-Arylimino-5,6-dihydro-4H-1,3-thiazines as a new class of cannabinoid receptor agonists. Part 1: Discovery of CB2 receptor selective compounds . Bioorganic & Medicinal Chemistry Letters. 2007 , 17 (14), 4030-4034.