Thermo Scientific Chemicals

2',3'-Dideoxycytidine, 98+%

Catalog number: L10619.MF
50 mg, Each
Thermo Scientific Chemicals

2',3'-Dideoxycytidine, 98+%

Catalog number: L10619.MF
50 mg, Each
Quantity
Catalog number: L10619.MF
also known as L10619-MF
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7481-89-2
IUPAC Name
4-amino-1-[(2R,5S)-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one
Molecular Formula
C9H13N3O3
InChI Key
WREGKURFCTUGRC-KGQMAECUNA-N
SMILES
NC1=NC(=O)N(C=C1)[C@H]1CC[C@@H](CO)O1
Appearance (Color)
White to pale cream
Form
Crystals or crystalline powder or powder
Assay (HPLC)
≥98.0%
Identification (FTIR)
Conforms

Description

2',3'-Dideoxycytidine in cells, it is phosphorylated to the active triphosphate form, ddCTP, which acts as a substrate for HIV reverse transcriptase (Ki = 51 nM). It incorporates into viral DNA where it terminates chain elongation when the missing hydroxyl group is encountered. It is also is used as a DNA chain-terminating nucleotide for DNA sequencing methods based on the Sanger chain-termination method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text