3-Methylquinoline N-oxide, 97%, Thermo Scientific Chemicals
3-Methylquinoline N-oxide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Methylquinoline N-oxide, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 g
Catalog number L10635.03
also known as L10635-03
Price (USD)/ Each
117.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
117.00
Add to cart
3-Methylquinoline N-oxide, 97%, Thermo Scientific Chemicals
Catalog numberL10635.03
Price (USD)/ Each
117.00
-
Add to cart
Chemical Identifiers
CAS1873-55-8
IUPAC Name3-methylquinolin-1-ium-1-olate
Molecular FormulaC10H9NO
InChI KeyCBGUMTBCWWMBRV-UHFFFAOYSA-N
SMILESCC1=C[N+]([O-])=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to pale brown
Assay (HPLC)≥96.0%
Melting Point (clear melt)83.5-92.5?C
FormCrystals or powder or crystalline powder or granules
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only