N-Isopropylaniline, 98%, Thermo Scientific Chemicals
N-Isopropylaniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Isopropylaniline, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number L10646.18
also known as L10646-18
Price (USD)/ Each
150.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
150.00
Add to cart
N-Isopropylaniline, 98%, Thermo Scientific Chemicals
Catalog numberL10646.18
Price (USD)/ Each
150.00
-
Add to cart
Chemical Identifiers
CAS768-52-5
IUPAC NameN-(propan-2-yl)aniline
Molecular FormulaC9H13N
InChI KeyFRCFWPVMFJMNDP-UHFFFAOYSA-N
SMILESCC(C)NC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to dark yellow to orange
Assay (GC)≥97.5%
Refractive Index1.5365-1.5405 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only