Thermo Scientific Chemicals

2,6-Dibromo-4-(trifluoromethyl)aniline, 97%, Thermo Scientific Chemicals

Catalog number: L10658.03
1 g, Each
Thermo Scientific Chemicals

2,6-Dibromo-4-(trifluoromethyl)aniline, 97%, Thermo Scientific Chemicals

Catalog number: L10658.03
1 g, Each
Quantity
Catalog number: L10658.03
also known as L10658-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
72678-19-4
IUPAC Name
2,6-dibromo-4-(trifluoromethyl)aniline
Molecular Formula
C7H4Br2F3N
InChI Key
DRSMEHXBOXHXDX-UHFFFAOYSA-N
SMILES
NC1=C(Br)C=C(C=C1Br)C(F)(F)F
Appearance (Color)
White to cream or pale yellow
Assay (GC)
≥96.0%
Melting Point (clear melt)
36-41?C
Form
Crystals or crystalline powder of fused solid

Description

Used in enantio-and diastereoselective addition reation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in enantio-and diastereoselective addition reation.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text