Thermo Scientific Chemicals

trans-5-Decene, 97%, Thermo Scientific Chemicals

Catalog number: L10689.06
5 g, Each
Thermo Scientific Chemicals

trans-5-Decene, 97%, Thermo Scientific Chemicals

Catalog number: L10689.06
5 g, Each
Quantity
Catalog number: L10689.06
also known as L10689-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7433-56-9
Assay (GC)
≥96.0%
Refractive Index
1.4210-1.4260 @ 20°C (non-U.S. sourced material)
Appearance (Color)
Clear colorless
Form
Liquid
Comment
Material sourced in the U.S. and in other countries

Description

trans-5-Decene has been used to characterize the effect of cyclization and size on the stability of Criegee intermediates formed in ozonolysis. It has been added to phenoxathiin cation radical in acetonitrile solution to form bis(10-phenoxathiiniumyl) alkane adducts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-5-Decene has been used to characterize the effect of cyclization and size on the stability of Criegee intermediates formed in ozonolysis. It has been added to phenoxathiin cation radical in acetonitrile solution to form bis(10-phenoxathiiniumyl) alkane adducts.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text