Thermo Scientific Chemicals

2-Indanylacetic acid, 99%, Thermo Scientific Chemicals

Catalog number: L10700.03
1 g, Each
Thermo Scientific Chemicals

2-Indanylacetic acid, 99%, Thermo Scientific Chemicals

Catalog number: L10700.03
1 g, Each
Quantity
Catalog number: L10700.03
also known as L10700-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
37868-26-1
Appearance (Color)
White to cream to brown
Form
Crystals or powder or crystalline powder or fused solid
Assay (Silylated GC)
≥98.5%
Melting Point (clear melt)
88.0-95.0?C
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%

Description

2-Indanylacetic acid is the starting material in the synthesis of pentacylic core of (+)-Salvileucalin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Indanylacetic acid is the starting material in the synthesis of pentacylic core of (+)-Salvileucalin.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text