1-Chloro-2-iodoethane is used as a mild electrophile for iodination of metallated species, as in the conversion of an ortho-lithiated benzyl ether to the corresponding ortho-iodide. It reacts with 5-hydroxy-[1,4]naphthoquinone, and obtain the 5-(2-chloro-ethoxy)-[1,4]naphthoquinone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Chloro-2-iodoethane is used as a mild electrophile for iodination of metallated species, as in the conversion of an ortho-lithiated benzyl ether to the corresponding ortho-iodide. It reacts with 5-hydroxy-[1,4]naphthoquinone, and obtain the 5-(2-chloro-ethoxy)-[1,4]naphthoquinone.
Solubility
Soluble inorganic solvents. Insoluble in water.
Notes
Incompatible with strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
Xuming Zheng.; David Lee Phillips. A-band resonance Raman spectra and short-time photodissociation dynamics of trans 1-chloro-2-iodoethane in cyclohexane solution. Chemical Physics Letters. 1998, 286 (1-2), 79-87.
Xuming Zheng.; David Lee Phillips. Investigation of the short-time photodissociation dynamics of trans-1-bromo-2-iodoethane in the A -band absorption. J. Chem. Phys. 1999, 110 1638 .
Mild electrophile for iodination of metallated species, as in the conversion of an ortho-lithiated benzyl ether to the corresponding ortho-iodide: J. Chem. Soc., Chem. Commun., 124 (1979); J. Org. Chem., 47, 2541 (1982); J. Chem. Soc., Perkin 1, 115 (1985). See also 1,2-Diiodoethane, L03019.