Thermo Scientific Chemicals

1-Chloro-2-iodoethane, 96%, stab. with copper, Thermo Scientific Chemicals

Catalog number: L10742.14
25 g, Each
Thermo Scientific Chemicals

1-Chloro-2-iodoethane, 96%, stab. with copper, Thermo Scientific Chemicals

Catalog number: L10742.14
25 g, Each
Quantity
Catalog number: L10742.14
also known as L10742-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
624-70-4
IUPAC Name
1-chloro-2-iodoethane
Molecular Formula
C2H4ClI
InChI Key
JTWWWQGSFTWWDL-UHFFFAOYSA-N
SMILES
ClCCI
Appearance (Color)
Clear colorless to dark red/brown
Form
Liquid
Assay (GC)
>95.0%
Refractive Index
1.5615-1.5675 @ 20?C

Description

1-Chloro-2-iodoethane is used as a mild electrophile for iodination of metallated species, as in the conversion of an ortho-lithiated benzyl ether to the corresponding ortho-iodide. It reacts with 5-hydroxy-[1,4]naphthoquinone, and obtain the 5-(2-chloro-ethoxy)-[1,4]naphthoquinone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-2-iodoethane is used as a mild electrophile for iodination of metallated species, as in the conversion of an ortho-lithiated benzyl ether to the corresponding ortho-iodide. It reacts with 5-hydroxy-[1,4]naphthoquinone, and obtain the 5-(2-chloro-ethoxy)-[1,4]naphthoquinone.

Solubility
Soluble inorganic solvents. Insoluble in water.

Notes
Incompatible with strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text