Thermo Scientific Chemicals

Vinylcyclohexane, 97%, Thermo Scientific Chemicals

Catalog number: L10756.06
5 g, Each
Thermo Scientific Chemicals

Vinylcyclohexane, 97%, Thermo Scientific Chemicals

Catalog number: L10756.06
5 g, Each
Quantity
Catalog number: L10756.06
also known as L10756-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
695-12-5
IUPAC Name
ethenylcyclohexane
Molecular Formula
C8H14
InChI Key
LDLDYFCCDKENPD-UHFFFAOYSA-N
SMILES
C=CC1CCCCC1
Solution Test
10% in methanol: clear
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≤96.0%
Identification (FTIR)
Conforms

Description

The use of vinylcyclohexane dioxide (VCD) as a polar dehydrant with subsequent embedment in Spurr was studied. Vinylcyclohexane (VCH) was copolymerized with ethene and propene using methylaluminoxane-activated metallocene catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The use of vinylcyclohexane dioxide (VCD) as a polar dehydrant with subsequent embedment in Spurr was studied. Vinylcyclohexane (VCH) was copolymerized with ethene and propene using methylaluminoxane-activated metallocene catalysts.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text