Thermo Scientific Chemicals

Propiolaldehyde diethyl acetal, 97%, Thermo Scientific Chemicals

Catalog number: L10767.03
1 g, Each
Thermo Scientific Chemicals

Propiolaldehyde diethyl acetal, 97%, Thermo Scientific Chemicals

Catalog number: L10767.03
1 g, Each
Quantity
Catalog number: L10767.03
also known as L10767-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10160-87-9
IUPAC Name
3,3-diethoxyprop-1-yne
Molecular Formula
C7H12O2
InChI Key
RGUXEWWHSQGVRZ-UHFFFAOYSA-N
SMILES
CCOC(OCC)C#C
Form
Liquid
Assay (GC)
≥96.0%
Refractive Index
1.4100-1.4150 @ 20°C (non-U.S. sourced material)
Appearance (Color)
Clear colorless to yellow
Comment
Material sourced in the U.S. and in other countries

Description

Propiolaldehyde diethyl acetal is used in preparation of 3-boronoacrolein pinacolate, a heterodyne and (E)-3-(tributylstannyl)-2-propenal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Propiolaldehyde diethyl acetal is used in preparation of 3-boronoacrolein pinacolate, a heterodyne and (E)-3-(tributylstannyl)-2-propenal.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. Air and moisture sensitive. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text