Thermo Scientific Chemicals

3-Hydroxy-1-methylpiperidine, 98%, Thermo Scientific Chemicals

Catalog number: L10829.06
5 g, Each
Thermo Scientific Chemicals

3-Hydroxy-1-methylpiperidine, 98%, Thermo Scientific Chemicals

Catalog number: L10829.06
5 g, Each
Quantity
Catalog number: L10829.06
also known as L10829-06
Price (USD)
Have Questions?

Chemical Identifiers

CAS
3554-74-3
IUPAC Name
1-methylpiperidin-3-ol
Molecular Formula
C6H13NO
InChI Key
UKANCZCEGQDKGF-UHFFFAOYNA-N
SMILES
CN1CCCC(O)C1
Form
Liquid or viscous liquid
Appearance (Color)
Clear colorless to orange
Assay (GC)
≥97.5%
Refractive Index
1.4720-1.4760 @ 20?C

Description

Reactant for Optimization of Novobiocin scaffold to produce antitumor agents, Formation of carbamates and N-alkylimidazoles. Reactant for synthesis of Pyridine derivatives as CDK5 inhibitors Phenylcarbamate derivatives as ligands for nicotinic acetylcholine receptors, Amino phosphite ligands, Mexiletine enantiomers by nucleophilic aromatic substitution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant for Optimization of Novobiocin scaffold to produce antitumor agents, Formation of carbamates and N-alkylimidazoles. Reactant for synthesis of Pyridine derivatives as CDK5 inhibitors Phenylcarbamate derivatives as ligands for nicotinic acetylcholine receptors, Amino phosphite ligands, Mexiletine enantiomers by nucleophilic aromatic substitution.

Solubility
Fully misicible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text