Thermo Scientific Chemicals

4-Bromophenyl isocyanate, 99%, Thermo Scientific Chemicals

Catalog number: L10834.14
25 g, Each
Thermo Scientific Chemicals

4-Bromophenyl isocyanate, 99%, Thermo Scientific Chemicals

Catalog number: L10834.14
25 g, Each
Quantity
Catalog number: L10834.14
also known as L10834-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2493-02-9
IUPAC Name
1-bromo-4-isocyanatobenzene
Molecular Formula
C7H4BrNO
InChI Key
CZQIJQFTRGDODI-UHFFFAOYSA-N
SMILES
BrC1=CC=C(C=C1)N=C=O
Appearance (Color)
White to pale cream
Identification (FTIR)
Conforms
Melting Point (clear melt)
38.0-44.0?C
Form
Crystals or powder or crystalline powder or lumps or fused solid
Assay (GC)
≥98.5%

Description

4-Bromophenyl isocyanate is used in the synthesis of mixed bisamide derivative and monoamide product.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromophenyl isocyanate is used in the synthesis of mixed bisamide derivative and monoamide product.

Solubility
Sparingly soluble in water (0.40 g/L at 25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text