Benzyl methacrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Benzyl methacrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to light and heat. Incompatible with oxidizing agents.
RUO – Research Use Only
Mona Semsarilar.; Elizabeth R. Jones.; Adam Blanazs.; Steven P. Armes. Efficient Synthesis of Sterically-Stabilized Nano-Objects via RAFT Dispersion Polymerization of Benzyl Methacrylate in Alcoholic Media. Advanced Materials. 2012, 24 (25), 3378-3382.
John Mykytiuk.; Steven P. Armes.; Norman C. Billingham. Group-transfer polymerization of benzyl methacrylate: A convenient method for synthesis of near-monodisperse poly(methacrylic acid)s. Polymer Bulletin. 1992, 29 (1), 139-145.