Methyl 3,5-diaminobenzoate, 99%, Thermo Scientific Chemicals
Methyl 3,5-diaminobenzoate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 3,5-diaminobenzoate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number L10848.18
also known as L10848-18
Price (USD)/ Each
249.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
249.00
Add to cart
Methyl 3,5-diaminobenzoate, 99%, Thermo Scientific Chemicals
Catalog numberL10848.18
Price (USD)/ Each
249.00
-
Add to cart
Chemical Identifiers
CAS1949-55-9
IUPAC Namemethyl 3,5-diaminobenzoate
Molecular FormulaC8H10N2O2
InChI KeyIYWLDOMAEQHKJZ-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC(N)=CC(N)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)130.0-136.0?C
Appearance (Color)Cream to yellow to brown
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥98.5%
Methyl 3,5-diaminobenzoate is used as a primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3,5-diaminobenzoate is used as a primary and secondary intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong bases, strong acids and strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Hak-Fun Chow.; Jie Zhang.; Chui-Man Lo.; Siu-Yin Cheung.; Ka-Wai Wong. Improving the gelation properties of 3,5-diaminobenzoate-based organogelators in aromatic solvents with additional aromatic-containing pendants. Tetrahedron. 2007, 63 (2), 363-373.
  2. P. Kimkes.; U. Sohling.; G. T. Oostergetel.; A. J. Schouten. Oxidative Coupling Polymerization in a Langmuir Monolayer of Octadecyl 3,5-Diaminobenzoate. Langmuir. 1996, 12 (16), 3945-3951.