n-Pentyl acetate, 99%, Thermo Scientific Chemicals
n-Pentyl acetate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

n-Pentyl acetate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number L10852.22
also known as L10852-22
Price (USD)/ Each
26.10
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
26.10
Add to cart
n-Pentyl acetate, 99%, Thermo Scientific Chemicals
Catalog numberL10852.22
Price (USD)/ Each
26.10
-
Add to cart
Chemical Identifiers
CAS628-63-7
IUPAC Namepentyl acetate
Molecular FormulaC7H14O2
InChI KeyPGMYKACGEOXYJE-UHFFFAOYSA-N
SMILESCCCCCOC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Refractive Index1.4005-1.4035 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
FormLiquid
n-Pentyl acetate is used as in studies of olfactory function and in studies of the psychosocial effects of odor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Pentyl acetate is used as in studies of olfactory function and in studies of the psychosocial effects of odor.

Solubility
Slightly soluble in water (2g/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong bases and strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Weixian Chang.; Guofeng Guan.; Xinli Li.; Huqing Yao. Isobaric Vapor-Liquid Equilibria for Water + Acetic Acid + (n-Pentyl Acetate or Isopropyl Acetate). J. Chem. Eng. Data. 2005, 50 (4), 1129-1133.
  2. B.M.S Santos.; A.G.M Ferreira.; I.M.A Fonseca. Surface and interfacial tensions of the systems water + n-butyl acetate + methanol and water + n-pentyl acetate + methanol at 303.15 K. Fluid Phase Equilibria. 2003, 208 (1-2), 1-21.