Thermo Scientific Chemicals

n-Pentyl acetate, 99%, Thermo Scientific Chemicals

Catalog number: L10852.36
500 g, Each
Thermo Scientific Chemicals

n-Pentyl acetate, 99%, Thermo Scientific Chemicals

Catalog number: L10852.36
500 g, Each
Quantity
Catalog number: L10852.36
also known as L10852-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
628-63-7
IUPAC Name
pentyl acetate
Molecular Formula
C7H14O2
InChI Key
PGMYKACGEOXYJE-UHFFFAOYSA-N
SMILES
CCCCCOC(C)=O
Form
Liquid
Identification (FTIR)
Conforms
Refractive Index
1.4005-1.4035 @ 20?C
Appearance (Color)
Clear colorless
Assay (GC)
≥98.5%

Description

n-Pentyl acetate is used as in studies of olfactory function and in studies of the psychosocial effects of odor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Pentyl acetate is used as in studies of olfactory function and in studies of the psychosocial effects of odor.

Solubility
Slightly soluble in water (2g/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong bases and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text